Drukuj

Anti-GNAO1 Rabbit Polyclonal Antibody (Cy5®)

Dostawca: Bioss
BOSSBS-6962R-CY5EA 4310 PLN
BOSSBS-6962R-CY5
Anti-GNAO1 Rabbit Polyclonal Antibody (Cy5®)
Przeciwciała
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems. The G(o) protein function is not clear. Stimulated by RGS14.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNAO1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR