Drukuj
Q-BioTaq™ CORE KIT 10
Dostawca: MP Biomedicals
ICNA11EPQBK1025EA 1060 PLN
ICNA11EPQBK1025
Q-BioTaq™ CORE KIT 10
Odczynniki do kwasów nukleinowych End-point PCR Enzymes and Kits
Q-BioTaq™ CORE KIT 10 is used to amplify DNA sequences. It is particularly useful in applications where multiple targets are amplified (multiplexing).
Q-BioTaq™ DNA polymerase is a thermostable DNA polymerase which can be used effectively for PCR and DNA sequencing by the chain termination method. This enzyme is encoded by a modified form of the Thermus aquaticus DNA polymerase gene with an N-terminal deletion. The properties of QBioTaq™ DNA polymerase include high thermostability (98 °C) and absence of 5’-3’ exonuclease activity. The enzyme is highly purified and is free of endo- and exonucleases.
Informacje dodatkowe: Kit contains all the reagents required for PCR including: Q-BioTaq™ DNA polymerase, dNTP Mix (>99% purity), optimised incubation buffers with and without MgCl₂ and a separate vial of MgCl₂.