Order Entry
Pipette holder, SINGLE™
  GILSF161406
 :  GILSON
undefined
Pipette holder, SINGLE™
  GILSF161406
 :  GILSON
92 zł
  1 * 1 SZT
  • Colour:
    White
  • Description:
    Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws
  • Pk:
    1
This convenient pipette holder has been designed to hold one single or multi channel pipette.
  • Made of PC: Resistant to aggressive chemicals and UV light
  • Adheres strongly to any smooth, dry surface or may be fastened with screws
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Space saving: Adapted to limited work bench space