Drukuj
Peanut butter, NIST-2387 standard
Dostawca: LGC Standards
PROONIST-2387EA 8500 PLN
PROONIST-2387
Peanut butter, NIST-2387 standard
Standardy Wzorce żywności
This reference material intended to use in validating methods for determining fat, fatty acids, trace elements and tocopherol.
Certyfikaty: Traceable to NIST and SRM.