Capillary bottle, CERTAN®
Supplier: Merck
Certificates
About this item
Capillary bottles with PTFE-lined screw-caps are designed for the storage of volatile samples and reference standards.
- No appreciable change in concentration
- Secure storage, minimal evaporation
- Minimizes contamination risk
- Sample filling and removal using a standard GC syringe
- Essentially spillproof
It is applicable for storing liquids removed from conventional glass ampules, stock and standard solutions, difficult samples such as volatile halogen compounds or BTEX aromatics and archiving of production or QC samples and extracts.
Product Information
- Catalog #Supplier #CapacityØ×HCap includedPkAvailabilityPrice
- Capillary bottleCapillary bottleSIAL44419-10EA44419-10EA1,50 ml12×50 mmYes1Log in for availability1 * 1 Szt.724 złSupplier Article Number: 44419-10EALocal Article Number: SIAL44419-10EASpecifications:
Specifications
Environmentally Preferable - Capillary bottleCapillary bottleSIAL44420-5EA44420-5EA4,50 ml16×71 mmYes1Log in for availability1 * 1 Szt.431 złSupplier Article Number: 44420-5EALocal Article Number: SIAL44420-5EASpecifications:
Specifications
Environmentally Preferable - Capillary bottleCapillary bottleSIAL44421-5EA44421-5EA10,00 ml30×71 mmYes1Log in for availability1 * 1 Szt.495 złSupplier Article Number: 44421-5EALocal Article Number: SIAL44421-5EASpecifications:
Specifications
Environmentally Preferable