Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-HPRT1 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
HPRT1 has a central role in the generation of purine nucleotides through the purine salvage pathway. HPRT1 catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate.HPRT1 has a central role in the generation of purine nucleotides through the purine salvage pathway. HPRT1 catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate (Keebaugh et al., 2007 [PubMed 16928426]).

HPRT1 antibody can be used for detection of HPRT1 by ELISA at 1:12500. HPRT1 antibody can be used for detection of HPRT1 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HPRT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
50 µg 50 µG PRSI26-316
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol HPRT1
Antigen name Hypoxanthine phosphoribosyltransferase 1
Reactivity Mouse, Human, Rat, Dog
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms HGPRT, Hypoxanthine-guanine phosphoribosyltransferase, HGPRTase, HPRT, HPRT1
Immunogen A synthetic peptide corresponding a region of human HPRT1.
Form Lyophilized
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość