Drukuj

Anti-NAA30 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
PRSI26-320EA 2960 PLN
PRSI26-320
Anti-NAA30 Rabbit Polyclonal Antibody
Przeciwciała
NAT12 belongs to the acetyltransferase family, MAK3 subfamily. It contains 1 N-acetyltransferase domain. It is a probable N-acetyltransferase.

NAT12 antibody can be used for detection of NAT12 by ELISA at 1:62500. NAT12 antibody can be used for detection of NAT12 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NAA30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR