Drukuj

Anti-SULT1C4 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
PRSI26-339EA 2960 PLN
PRSI26-339
Anti-SULT1C4 Rabbit Polyclonal Antibody
Przeciwciała
SULT1C4 catalyzes the sulfate conjugation of many drugs, xenobiotic compounds, hormones, and neurotransmitters. It may be involved in the activation of carcinogenic hyroxylamines. SULT1C4 shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF).Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that belongs to the SULT1 subfamily, responsible for transferring a sulfo moiety from PAPS to phenol-containing compounds.

SULT1C4 antibody can be used for detection of SULT1C4 by ELISA at 1:62500. SULT1C4 antibody can be used for detection of SULT1C4 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SULT1C4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR