Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-POLK Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
POLK belongs to the DNA polymerase type-Y family. It contains 2 Rad18-type zinc fingers and 1 umuC domain. POLK is a DNA polymerase specifically involved in DNA repair. It plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Depending on the context, it inserts the correct base, but causes frequent base transitions, transversions and frameshifts. It lacks 3'-5' proofreading exonuclease activity. POLK forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but does not have lyase activity.

POLK antibody can be used for detection of POLK by ELISA at 1:62500. POLK antibody can be used for detection of POLK by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
50 µg 50 µG PRSI26-347
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol POLK
Antigen name Polymerase (DNA Directed) Kappa
Reactivity Human, Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms OTTHUMP00000020323, BETAN, RP11-529I10.1, DNA-directed DNA polymerase lambda, lambda, POLL, pol lambda, DNA polymerase kappa (DNA polymerase beta-N), pol beta2, polymerase (DNA-directed), DNA polymerase lamda2, polymerase (DNA directed), FLJ46002, DNA polymerase beta-2, BETA-N, POL-KAPPA, POLKAPPA
Immunogen A synthetic peptide corresponding a region of human POLK.
Form Lyophilized
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość