Drukuj

Anti-THUMPD2 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
PRSI26-363EA 2960 PLN
PRSI26-363
Anti-THUMPD2 Rabbit Polyclonal Antibody
Przeciwciała
THUMPD2 belongs to the methyltransferase superfamily. It contains 1 THUMP domain. The function of the THUMPD2 protein remains unknown.

THUMPD2 antibody can be used for detection of THUMPD2 by ELISA at 1:312500. THUMPD2 antibody can be used for detection of THUMPD2 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: THUMPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR