Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-B3GNT4 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
B3GNT4 is a member of the beta-1,3-N-acetylglucosaminyltransferase protein family. The protein is involved in the biosynthesis of poly-N-acetyllactosamine chains and prefers lacto-N-neotetraose as a substrate. It is a type II transmembrane protein.This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase protein family. The encoded enzyme is involved in the biosynthesis of poly-N-acetyllactosamine chains and prefers lacto-N-neotetraose as a substrate. It is a type II transmembrane protein.

B3GNT4 antibody can be used for detection of B3GNT4 by ELISA at 1:312500. B3GNT4 antibody can be used for detection of B3GNT4 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: B3GNT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
50 µg 50 µG PRSI26-364
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol B3GNT4
Antigen name betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human B3GNT4.
Form Lyophilized
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość