Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-METTL15 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
METT5D1 belongs to the methyltransferase superfamily, mraW family. METT5D1 is a probable S-adenosyl-L-methionine-dependent methyltransferase.

METT5D1 antibody can be used for detection of METT5D1 by ELISA at 1:1562500. METT5D1 antibody can be used for detection of METT5D1 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: METTL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
50 µg 50 µG PRSI26-374
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol METTL15
Antigen name Methyltransferase Like 15
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms METT5D1
Immunogen A synthetic peptide corresponding a region of human METT5D1.
Form Lyophilized
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość