Drukuj

Anti-GSTO2 Rabbit Polyclonal Antibody

Dostawca: ProSci Inc.
PRSI26-383EA 2960 PLN
PRSI26-383
Anti-GSTO2 Rabbit Polyclonal Antibody
Przeciwciała
The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca (2+) release by ryanodine receptors (e.g., RYR1).

GSTO2 antibody can be used for detection of GSTO2 by ELISA at 1:312500. GSTO2 antibody can be used for detection of GSTO2 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GSTO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR