Anti-PAX3 Goat Polyclonal Antibody
PRSI42-977
- Antigen name:Paired Box 3
- Antigen symbol:PAX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CDHS|paired box protein Pax-3|WS3|WS1|paired domain gene HuP2|paired domain gene 3|HUP2|paired box homeotic gene 3
- Concentration:500 ug/mL
- Immunogen:PAX3 antibody was raised against a 13 amino acid synthetic peptide near the internal region of PAX3.
- Size:100 µg
- Pk:100 µG
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PAX3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PAX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat