Drukuj
Human recombinant Tnfsf18 (from E. coli)
Dostawca: ProSci
PRSI40-313EA 968 PLN
PRSI40-313
Human recombinant Tnfsf18 (from E. coli)
AITRL, a member of the TNF superfamily, is expressed in endothelial cells, and signals through the AITR receptor. AITRL regulates T-cell proliferation and survival, and effectuates the interaction between T lymphocytes and endothelial cells. The AITRL gene codes for a type II transmembrane protein comprised of 177 amino acids, including a 28 amino acid cytoplasmic region, a 21 amino acid transmembrane domain and a 128 amino acid extracellular domain. Recombinant human soluble AITRL is a 14.3 kDa protein, containing 126 amino acid residues corresponding to the extracellular domain of AITRL.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.