Drukuj
Human Recombinant GITR Ligand (from Cells)
Dostawca: ProSci
PRSI92-315EA 2200 PLN
PRSI92-315
Human Recombinant GITR Ligand (from Cells)
Białka
Cytokine that binds to TNFRSF18/AITR/GITR. Regulates T-cell responses. Can function as costimulator and lower the threshold for T-cell activation and T-cell proliferation. Important for interactions between activated T-lymphocytes and endothelial cells. Mediates activation of NF-kappa-B.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.