Mono-Reactive NHS Ester dye packs, Amersham CyDye™
Supplier: Cytiva
Certificates
About this item
Specifications
- Emission max.:694 nm
Specifications
Related Information
Certificates
Search by Lot Number25000699_INSTRUCTIONS 25000699_INSTRUCTIONS 1 25005372_INSTRUCTIONS 25005372_INSTRUCTIONS 1 25005383_INSTRUCTIONS
Manual
More
25190143_DATA FILE 25190143_DATA FILE 1 25190149_DATA FILE 25190149_DATA FILE 1 25800305_DATA FILE
Datasheet
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Cy5,5 NHS Ester, 694 nmCy5,5 NHS Ester, 694 nmPA15601PA156011 mg1 * 1 mg1 870 złSupplier Article Number: PA15601Local Article Number: GEHEPA15601
- Cy5 NHS Ester, 670 nmCy5 NHS Ester, 670 nmPA15106PA151061 KIT1 * 1 KIT24 950 złSupplier Article Number: PA15106Local Article Number: GEHEPA15106
- Cy5 NHS Ester, 670 nmCy5 NHS Ester, 670 nmPA15104PA151041 KIT1 * 1 KIT15 080 złSupplier Article Number: PA15104Local Article Number: GEHEPA15104
- Cy5 NHS Ester, 670 nmCy5 NHS Ester, 670 nmPA15102PA151021 KIT1 * 1 KIT49 580 złSupplier Article Number: PA15102Local Article Number: GEHEPA15102
- Cy5 NHS Ester, 670 nmCy5 NHS Ester, 670 nmPA15101PA151011 mg1 * 1 mg2 090 złSupplier Article Number: PA15101Local Article Number: GEHEPA15101
- Cy5 NHS Ester, 670 nmCy5 NHS Ester, 670 nmPA15100PA151005 mg1 * 5 mg8 500 złSupplier Article Number: PA15100Local Article Number: GEHEPA15100
- Cy5,5 NHS Ester, 694 nmCy5,5 NHS Ester, 694 nmPA15606PA1560625 mg1 * 25 mg23 780 złSupplier Article Number: PA15606Local Article Number: GEHEPA15606
- Cy5,5 NHS Ester, 694 nmCy5,5 NHS Ester, 694 nmPA15605PA156055 mg1 * 5 mg6 770 złSupplier Article Number: PA15605Local Article Number: GEHEPA15605
- Cy7 NHS Ester, 767 nmCy7 NHS Ester, 767 nmPA17104PA171041 KIT1 * 1 KIT11 620 złSupplier Article Number: PA17104Local Article Number: GEHEPA17104
- Cy5,5 NHS Ester, 694 nmCy5,5 NHS Ester, 694 nmPA15604PA1560410 mg1 * 10 mg10 140 złSupplier Article Number: PA15604Local Article Number: GEHEPA15604
- Cy3 NHS Ester, 570 nmCy3 NHS Ester, 570 nmPA13105PA131051 KIT1 * 1 KIT7 830 złSupplier Article Number: PA13105Local Article Number: GEHEPA13105
- Cy5,5 NHS Ester, 694 nmCy5,5 NHS Ester, 694 nmPA15602PA1560250 mg1 * 50 mg41 020 złSupplier Article Number: PA15602Local Article Number: GEHEPA15602
- Cy3 NHS Ester, 570 nmCy3 NHS Ester, 570 nmPA13104PA131041 KIT1 * 1 KIT11 210 złSupplier Article Number: PA13104Local Article Number: GEHEPA13104
- Cy3 NHS Ester, 570 nmCy3 NHS Ester, 570 nmPA13101PA131011 mg1 * 1 mg1 890 złSupplier Article Number: PA13101Local Article Number: GEHEPA13101
- Cy7 NHS Ester, 767 nmCy7 NHS Ester, 767 nmPA17101PA171011 mg1 * 1 mg2 270 złSupplier Article Number: PA17101Local Article Number: GEHEPA17101