Order Entry
Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Certificates
Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pk Info
    Pk
    Availability
    Price
  • PROOVHG-PTEN-100
    VHG-PTEN-100
    Tellurium
    Te
    1000 μg/ml
    5% HNO₃
    100 ml
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 312 zł
    312 zł
    1 * 100 mL
       312 zł
    Supplier Article Number: VHG-PTEN-100
    Local Article Number: PROOVHG-PTEN-100
  • PROOVHG-PTEN-500
    VHG-PTEN-500
    Tellurium
    Te
    1000 μg/ml
    5% HNO₃
    500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 710 zł
    710 zł
    1 * 500 mL
       710 zł
    Supplier Article Number: VHG-PTEN-500
    Local Article Number: PROOVHG-PTEN-500