Drukuj
Worki do transportu próbek InterLAB
Dostawca: Coveris
InterLAB®
COVSITL-001EA 813 PLN
COVSITL-001
Worki do transportu próbek InterLAB
Worki Woreczki na próbki
InterLAB® is designed for blood sampling sectors; it combines the highest levels of leak resistance and feature a ‘blade-less’ easy open system to bring real benefits to multiple analytical processes. It is used as internal in hospitals and laboratories and external secondary packaging for the transportation of biological substances of category B (UN3373).
  • Leak resistant for 100% protection: This avoid contamination risks on nosocomial diseases
  • Unique sequential numbering for traceability
  • Peelable adhesive to attach document for a cost effective and safe identification of sample bag; no sample confusion
  • Easy to use, it gives a perfect seal with a fast adhesive closure and has an easy and quick opening, this unique opening system gives perfect leak resistance which could not be achieved with a pre-cut opening perforation
How it works: The polyethylene specimen bag features a removable adhesive release liner that secures the specimen in a liquid-tight pouch – giving a permanent tamper resistant closure. On the reverse of the bag is a removable second adhesive liner; remove this to expose a peelable adhesive used to safely secure the requisition document to the pouch for transportation to the lab. At the lab the requisition document easily peels from the sample bag.
Certyfikaty: Compliant to International and European standard or guideline for the transport of infectious substances (ADR2017/P650 WHO/HSE/GCR/2012.12 - ISO 15189).