VWR®, High Contrast Achromatic Objectives for VisiScope
Supplier: VWR Collection
Certificates
About this item
These entry-level lenses are ideal for brightfield, darkfield and simple polarization applications. They are specifically designed to achieve optimal contrast and thus maximise yield on an instrument intended for education on F.N. 18.
Product Information
- Catalog #MagnificationPkAvailabilityPrice
- Achromatic objectiveAchromatic objective630-16284×/0,1011 * 1 SZT118 złSpecifications:
Specifications
Environmentally Preferable - Achromatic objectiveAchromatic objective630-162910×/0,2511 * 1 SZT135 złSpecifications:
Specifications
Environmentally Preferable - Achromatic objectiveAchromatic objective630-163020×/0,4011 * 1 SZT219 złSpecifications:
Specifications
Environmentally Preferable - Achromatic objectiveAchromatic objective630-163140×/0,6511 * 1 SZT396 złSpecifications:
Specifications
Environmentally Preferable - Achromatic objectiveAchromatic objective630-163260×/0,8511 * 1 SZT578 złSpecifications:
Specifications
Environmentally Preferable - Achromatic objectiveAchromatic objective630-1633100×/1,2511 * 1 SZT549 złSpecifications:
Specifications
Environmentally Preferable