Order Entry
Hydrogen generators
Hydrogen generators
New Product
undefined
Hydrogen generators
New Product
Ultra high purity: >99,99995% H₂; 0,69 - 6,89 bar for GC and GC/MS carrier gas applications
  •  
    Type
    Flow rate
    W×D×H
    Pk
  • DHUN110H-MDCARRIER
    110H-MDCARRIER
    110H-MDCARRIER
    1100 ml/min
    342×470×456 mm
    1
    1 * 1 SZT
    1 * 1 SZT
    New Product
    Accessories
    Specifications
    Environmentally Preferable
    New Product
Maintenance kits
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 07
    Desiccant cartridge for H models
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 685 zł
    685 zł
  •  
    Seq
    Description
    Pk
  • 10
    24 month preventative maintenance kit, for H models
    1
    1 * 1 SZT
    1 * 1 SZT - 4 130 zł
    4 130 zł
  •  
    Seq
    Description
    Pk
    Price
  • 08
    6 month preventative maintenance kit, for H models
    1
    87,40
    1 * 1 SZT
    1 * 1 SZT - 797 zł
    797 zł
  •  
    Seq
    Description
    Pk
  • 09
    6 month preventative maintenance kit, for H-MD models
    1
    1 * 1 SZT
    1 * 1 SZT - 680 zł
    680 zł
  •  
    Seq
    Description
    Pk
  • 11
    24 month preventative maintenance kit, for H-MD models
    1 KIT
    1 * 1 KIT
    1 * 1 KIT - 2 220 zł
    2 220 zł