The 45-tube powder dispenser is used to dispense the required amount of powder in 45 tubes placed in a BioSpec tube holder. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
- Simple, fast and reliable
- High productivity improvement 60-150 x
- Easy cleaning with 70% ethanol or soap
- Sterilisable and not autoclavable
- Ergonomic use as it removes tedious repetitive tasks
- Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminium
* Customised capacity (µl)
Dodatkowe informacje: Supplied with rack, converter and flight case.