Drukuj
Powder dispensers for 45 tubes
IBSA45-1-1000 IBSA45-1-50 IBSA45-1-100 IBSA45-1-250 IBSA45-1-999
Powder dispensers for 45 tubes
The 45-tube powder dispenser is used to dispense the required amount of powder in 45 tubes placed in a BioSpec tube holder. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
  • Simple, fast and reliable
  • High productivity improvement 60-150 x
  • Easy cleaning with 70% ethanol or soap
  • Sterilisable and not autoclavable
  • Ergonomic use as it removes tedious repetitive tasks
  • Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminium
* Customised capacity (µl)
Dodatkowe informacje: Supplied with rack, converter and flight case.