Protease Inhibitor I aldehyde
Cell permeable inhibitor of the 26S proteasome (MCP; multicatalytic proteinase complex) in HT4 cells. Causes accumulation of ubiquitinylated proteins in neuronal cells. Prevents the activation of NF-κB in response to TNF-α or okadaic acid through inhibition of IκBα degradation.
White powder.
Soluble in 100% ethanol (30 mg/ml), DMSO (30 mg/ml) or methanol (50 mg/ml).
- Storage conditions:Freezer
- Protein synonyms:Z-IE(OtBu)AL-CHO|Z-Ile-Glu(OtBu)-Ala-Leu-CHO
- Protein/peptide name:Protease Inhibitor I aldehyde
- Purity:95% (by TLC)
- Molecular weight:618,8
- Molecular formula:C₃₂H₅₀N₄O₈
Product Information
- Pk
- : ALX260089M001: ENZOALX260089M001
- : ALX260089M005: ENZOALX260089M005