Drukuj
Inkubatory rolkowe, standardowe i CO2
Dostawca: Wheaton
WHEAW753684-L-E
Inkubatory rolkowe, standardowe i CO2
These incubators are an ideal space saving and economical alternative to a warm room for small production runs. Designed specifically for use with the Wheaton roller culture apparatus, they can also be used in combination with optional shelves to accommodate spinner flasks, rocker tables and other equipment. Units have a viewing window in the door and four internal electrical outlets for shakers, stirrers etc. Units have a painted steel exterior, the standard models have painted steel interiors and CO₂ models have stainless steel interiors. Their double walled construction provides temperature stability and is easy to clean.
  • Forced air circulation with digital temperature control gives temperature variation (spatial) of ±0,5 at 37 °C (without shelves)
  • CO₂ incubator has lightweight lifting ramps for rolling in roller culture apparatus
  • CO₂ range 0 to 20%, gas shuts off when door is opened or switch is turned off
  • Safety features include independent over-temperature safety protection
  • Digital indicator and keypad, settings are maintained if power is interrupted
Informacje do zamówienia: Dostarczane bez półek, które są dostępne osobno.
Uwaga: * RT = Ambient