These 96-well starter bead dispensers are used to improve the consistency in dispensing while speeding up the process over 37 times.
- Simple, fast and reliable operation
- Fills a plate in only eight seconds
- Sterile and accurate dispensing with <1% error rate
- Easy cleaning
- Exchangeable bead plates
These dispensers are manual tools that dispenses beads accurately. By filling the dispenser and pulling the slider, a specific amount of beads is dropped in the container which can be tubes, microtiter plates or vials.
Several bead sizes or amounts can be dispensed simply by exchanging the metal bead plate from the bead dispenser system. The acrylic bead plate determines the size/amount of beads to be dispensed.
Dodatkowe informacje: Includes one standard mesh plate.