Chicken native albumin (from egg)
: Merck
Specifications
- CAS No.:9006-59-1
- Protein/peptide type:Native
- Source:Egg
- Species:Chicken
- Storage conditions:Refrigerator
- Protein/peptide name:Albumin
Specifications
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- SIALA5378-1GA5378-1GGlass bottle for solids1 g1 * 1 g290 zł: A5378-1G: SIALA5378-1G
- : A5378-5G: SIALA5378-5G
- SIALA5378-10GA5378-10GGlass bottle for solids10 g1 * 10 g1 500 zł: A5378-10G: SIALA5378-10G
- SIALA5378-25GA5378-25GGlass bottle for solids25 g1 * 25 g2 980 zł: A5378-25G: SIALA5378-25G
- SIALA5378-50GA5378-50GGlass bottle for solids50 g1 * 50 g5 130 zł: A5378-50G: SIALA5378-50G