Drukuj
glin 1000 mg/l w rozcieńczonym kwasie azotowym (from Al(NO3)3.9H2O) ARISTAR® pojedynczy wzorzec do ICP
Dostawca: Avantor
VWR® ARISTAR®
455002CEA 605 PLN
455002C 455004E 455012E 455014G
glin 1000 mg/l w rozcieńczonym kwasie azotowym (from Al(NO3)3.9H2O) ARISTAR® pojedynczy wzorzec do ICP
Standardy Wzorce kationowe Wzorce glinu
Al(NO₃)₃.9H₂O in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certyfikaty: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Dodatkowe informacje: Supplied with Certificate of Analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022