Drukuj
Chrom 10.000 mg/l w roztworze kwasu azotowego (from (NH4)2Cr2O7) ARISTAR® pojedynczy wzorzec do ICP
455232XEA 631 PLN
VWRC455232X VWRC455234Q
Chrom 10.000 mg/l w roztworze kwasu azotowego (from (NH4)2Cr2O7) ARISTAR® pojedynczy wzorzec do ICP
(NH₄)₂Cr₂O₇ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022