Sulphuric acid 2.5 M H2SO4 (5N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Density:1.180 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-755-4
UN:2796
ADR:8,II
:GHS05
Specification Test Results
- Factor:0.995 - 1.005
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Product Information
- Katalog #Dostawca #Pack typePkAvailabilityPrice
- Supplier Article Number: 35348-1LLocal Article Number: HONC35348-1L
- Supplier Article Number: 35348-5L-VPLocal Article Number: HONC35348-5L-VP
- Supplier Article Number: 35348-10L-VPLocal Article Number: HONC35348-10L-VP