Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
(S)-(-)-2-Phenyl-1-propanol
(S)-(-)-2-Phenyl-1-propanol, Sigma-Aldrich®
(S)-(-)-2-Phenyl-1-propanol, Sigma-Aldrich®
(S)-(-)-2-Phenyl-1-propanol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
(S)-(-)-2-Phenyl-1-propanol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
37778-99-7
:
Formula:
C₉H₁₂O
MDL Number:
MFCD00145249
:
136.19 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
SIAL78929-1G
78929-1G
Glass bottle for solids
1 g
Log in
for availability
Log in
for availability
1 * 1 g
-
+
Inquire For Price
1 * 1 g
-
+
Inquire For Price
SIAL78929-5G
78929-5G
Glass bottle for solids
5 g
Log in
for availability
Log in
for availability
1 * 5 g
-
+
Inquire For Price
1 * 5 g
-
+
Inquire For Price