2-Naphthyl acetate
MOLEM35023409
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
: Acetic acid β-naphthyl ester, Ethanoic acid β-naphthyl ester, Acetic acid 2-naphthyl ester, β-Naphthyl acetate, Ethanoic acid 2-naphthyl ester
:
:186.21 g/mol
:310
:68…71 °C
:1.1108 g/cm³ (70 °C)
:Freezer
:216-194-7
:1523-11-1
:C₁₂H₁₀O₂
:MFCD00004060