Sebacoyl dichloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-19-3
:
Formula:C₁₀H₁₆Cl₂O₂
MDL Number:MFCD00000770
:239.14 g/mol
Boiling Pt:220 °C (1013 hPa)
Melting Pt:–5 °C
Density:1.12 g/cm³ (20 °C)
Flash Pt:165 °C
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:203-843-4
:GHS05
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 236365-10GLocal Article Number: SIAL236365-10G
- Supplier Article Number: 236365-50GLocal Article Number: SIAL236365-50G