Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6976-37-0
:
Formula:C₈H₁₉NO₅
MDL Number:MFCD00002853
:209.24 g/mol
Melting Pt:102…103 °C
:M-Clarity™ quality level = MQ400
Storage Temperature:Ambient
EINECS:230-237-7
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice