1-Methylcyclopentanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1462-03-9
:
Formula:C₆H₁₂O
MDL Number:MFCD00001364
:100.16 g/mol
Boiling Pt:135 °C (1013 hPa)
Melting Pt:32…33 °C
Density:0.905 g/cm³ (23.5 °C)
EINECS:215-963-4
ADR:4.1,III
:M-Clarity™ quality level = MQ100
UN:1325