Selenious acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7783-00-8
:
Formula:H₂O₃Se
MDL Number:MFCD00011331
Merck Index:14,08430
Boiling Pt:315 °C (1013 hPa)
Melting Pt:70 °C
Density:1.998 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-974-7
UN:3283
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:128.97 g/mol
:GHS06
:GHS08
:GHS09