Ammonium sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7783-20-2
:
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391
:132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
Product Information
- Katalog #Dostawca #Pack typePkAvailabilityPrice
- Supplier Article Number: 204501-50GLocal Article Number: SIAL204501-50G
- Supplier Article Number: 204501-250GLocal Article Number: SIAL204501-250G