Materiały eksploatacyjne do ICP-MS Agilent Instruments serii 7700
Supplier: Agilent
Certificates
About this item
Cones and supplies
Product Information
- Catalog #Supplier #OpisDoIlośćAvailabilityPrice
- HEWLG3280-67063G3280-67063Platinum skimmer, Ni base7700x/e1Log in for availability1 * 1 SZT17 960 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67061G3280-67061Ni plated sampling cone7700 series1Log in for availability1 * 1 SZT6 840 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67064G3280-67064Platinum skimmer, Cu base7700s1Log in for availability1 * 1 SZT18 600 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67056G3280-67056Platinum (Pt) sampling cone, 18 mm insert, ICP-MS, for aqua regia digests, use with Pt skimmer cone and brass skimmer base7700/8800 series1Log in for availability1 * 1 SZT73 620 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67066G3280-67066Nickel (Ni) skimmer cone used with s-lens7700s and 8800 semiconductor ICP-MS1Log in for availability1 * 1 SZT4 550 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-60381G3280-60381Offset skimmer assembly7700s and 8800 semiconductor ICP-MS1Log in for availability1 * 1 SZT10 990 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67065G3280-67065Platinum (Pt) skimmer cone (Ni base) used with s-lens7700s and 8800 semiconductor ICP-MS1Log in for availability1 * 1 SZT18 600 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67036G3280-67036Platinum sampling cone7700s, 7700x/e1Log in for availability1 * 1 SZT27 070 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67009G3280-67009Graphite gasket for sampling cone7700x/e3Log in for availability1 * 3 SZT590 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-67060G3280-67060Platinum (Pt) skimmer cone used with x-lens, for the lowest limits of detection and for higher matrix samples, such as hydrofluoric acid digested geological samples7700x/e and 8800 series ICP-MS1Log in for availability1 * 1 SZT18 170 złSpecifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-60621G3280-60621Skimmer base, brass, for x-lens, use with Ni and Pt skimmer cones, includes screws7700x/e, but also suitable for 8800 ICP-MS1Log in for availability1 * 1 SZT4 690 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-60608G3280-60608Skimmer base, stainless steel, for x-lens, standard base, use with Ni sampling and skimmer cone. Includes screws7700x/e / 8800 ICP-MS1Log in for availability1 * 1 SZT4 740 złSpecifications:
Accessories
Specifications
Environmentally Preferable
Spray chamber supplies
Product Information
- Catalog #Supplier #OpisDoIlośćAvailabilityPrice
- HEWL5042-47755042-4775Connector for gas line at end cap Agilent ICP-MS7700 series1Log in for availability1 * 1 SZT349 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG8400-67026G8400-67026Dilution nozzle for ultra high matrix introduction, for Agilent 7900 ICP-MS7700 series1Log in for availability1 * 1 SZT3 300 złSpecifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - HEWLG8400-40200G8400-40200End cap retaining ring for spray chamber, for Agilent 7900 ICP-MS7700 series1Log in for availability1 * 1 SZT175 złSpecifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-60555G3280-60555Tubing assembly, drain, for ICP-MS7700 series1Log in for availability1 * 1 SZT1 630 złSpecifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - HEWLG1833-65426G1833-65426Connector between spray chamber and torch (quartz)7700 series1Log in for availability1 * 1 SZTSpecifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - HEWLG1833-65411G1833-65411Drain tubing kit (tubing between spray chamber and peri-pump, includes two connectors, bushing and tubing)7700 series1Log in for availability1 * 1 SZTSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG8400-67150G8400-67150Spray chamber (quartz) for UHMI, 7900 ICP-MS7700 series1Log in for availability1 * 1 SZT6 910 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG1833-65430G1833-65430Clamp for spray chamber, to clamp spray chamber/connector/torch7700 series1 KITLog in for availability1 * 1 KIT664 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG1820-65016G1820-65016Drain Tank (with PVC connector, 1 L)7700 series1Log in for availability1 * 1 Szt.1 120 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-80603G3280-80603Spray chamber, single-pass, for capillary LC, suits capillary LC interface kit used with Agilent 7700/8800 series ICP-MS7700 series1Log in for availability1 * 1 SZT10 320 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3280-80503G3280-80503Spray chamber, special long arm, for Agilent 7700/8000 series ICP-MS, recommended for ultra-trace analysis of B7700 series1Log in for availability1 * 1 SZT13 310 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG1820-65505G1820-65505Tank, drain, for organic solvent, PP connector, 2 L7700 series1Log in for availability1 * 1 SZT1 290 złSpecifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable
Nebuliser and supplies
Product Information
- Catalog #Supplier #OpisDoIlośćAvailabilityPrice
- HEWL0535-10820535-1082End cap nut for ICO-MS nebuliser connection7700 series1Log in for availability1 * 1 SZT141 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3139-65102G3139-65102MicroFlow nebuliser with I-AS probe7700 series1Log in for availability1 * 1 SZT18 170 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3266-80010G3266-80010Sample tubing for MicroMist, 0,25 mm id7700 series10Log in for availability1 * 10 SZT2 420 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3139-65100G3139-65100MicroFlow nebuliser7700 series1Log in for availability1 * 1 SZT16 030 złSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG3266-80011G3266-80011Sample tubing for MicroMist, 0,5 mm I.Ø7700 series10Log in for availability1 * 10 SZT2 040 złSpecifications:
Accessories
Specifications
Environmentally Preferable
Nebuliser and supplies
Product Information
- Catalog #OpisIlośćAvailabilityPrice
Vacuum system supplies
Product Information
- Catalog #SeqOpisDoIlośćCenaAvailabilityPrice