Hydrophobic interaction chromatography columns, HiTrap™ Capto™ Butyl (High Sub)
: Cytiva
Capto™ Phenyl (High sub) and Capto™ Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
- Aliphatic ligand coupled with a flexible spacer for laboratory and process applications requiring a HIC resin with low to resin hydrophobicity
- Highly rigid matrix enables high flow rates, long bed heights and faster throughput
- Improves productivity and process economy in downstream operations
- Narrow specification range ensures robust performance in sensitive processes
- Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide
- Catalog No:
- GEHE17-5459-08
- GEHE17-5459-09
- Storage:
- 4 to 30°C, 20% Ethanol
- 4 to 30°C, 20% Ethanol
- Column hardwareBed dimensionsCapacityFlow velocityPk
- HiTrap™ Capto™ Butyl (High Sub)HiTrap™ Capto™ Butyl (High Sub)GEHE17-5459-0817-5459-08PP7×25 mm1 ml<600 cm/h51 * 5 SZT1 400 zł
Specifications
Environmentally PreferableStorage4 to 30°C, 20% Ethanol - HiTrap™ Capto™ Butyl (High Sub)HiTrap™ Capto™ Butyl (High Sub)GEHE17-5459-0917-5459-09PP16×25 mm5 ml<600 cm/h51 * 5 SZT3 040 zł
Specifications
Environmentally PreferableStorage4 to 30°C, 20% Ethanol