Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
Electrophoresis Reagents
Molecular Weight Markers
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
SM0271
:
Thermo Fisher Scientific
:
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
SM0271
:
Thermo Fisher Scientific
:
380 zł
1 * 1 KIT
-
+
Pk:
1 KIT
Description:
pBR322 DNA/BsuRI (HaeIII) marker, 50 µg