ASTM® Precision Thermometers
Supplier: Amarell
Certificates
About this item
These models are suitable for government testing or official calibration.
- Solid stem thermometer
- White backed
Certifications: Compliant with ASTM.
With Work Certificate
Product Information
- Catalog #Supplier #RangeDivisionLengthFillingImmersion depthAvailabilityPrice
- ASTM precision thermometer, ASTM 1 CASTM precision thermometer, ASTM 1 CAMARA300010-CB/WCA300010-CB/WC−20...+150 °C1,0 °C322 mmBlue special liquid76 mmLog in for availability1 * 1 SZT1 260 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 1 CASTM precision thermometer, ASTM 1 CAMARA300010-FL/WCA300010-FL/WC−20...+150 °C1,0 °C322 mmRed special liquid76 mmLog in for availability1 * 1 SZT1 030 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 2 CASTM precision thermometer, ASTM 2 CAMARA300030-GA/WCA300030-GA/WC−5...+300 °C1,0 °C390 mmGallium76 mmLog in for availability1 * 1 SZT1 630 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 3 CASTM precision thermometer, ASTM 3 CAMARA300050-GA/WCA300050-GA/WC−5...+400 °C1,0 °C415 mmGallium76 mmLog in for availability1 * 1 SZT1 840 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 5 CASTM precision thermometer, ASTM 5 CAMARA300070-CB/WCA300070-CB/WC−38...+50 °C1,0 °C230 mmBlue special liquid108 mmLog in for availability1 * 1 SZT1 180 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 5 CASTM precision thermometer, ASTM 5 CAMARA300070-FL/WCA300070-FL/WC−38...+50 °C1,0 °C230 mmRed special liquid108 mmLog in for availability1 * 1 SZT948 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 6 CASTM precision thermometer, ASTM 6 CAMARA300090-TOL/WCA300090-TOL/WC−80...+20 °C1,0 °C230 mmToluene76 mmLog in for availability1 * 1 SZT1 070 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 7 CASTM precision thermometer, ASTM 7 CAMARA300110-GA/WCA300110-GA/WC−2...+300 °C1,0 °C385 mmGalliumTotalLog in for availability1 * 1 SZT1 710 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 8 CASTM precision thermometer, ASTM 8 CAMARA300130-GA/WCA300130-GA/WC−2...+400 °C1,0 °C385 mmGalliumTotalLog in for availability1 * 1 SZT1 660 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 9 CASTM precision thermometer, ASTM 9 CAMARA300150-CB/WCA300150-CB/WC−5...+110 °C0,5 °C290 mmBlue special liquid57 mmLog in for availability1 * 1 SZT1 240 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 9 CASTM precision thermometer, ASTM 9 CAMARA300150-FL/WCA300150-FL/WC−5...+110 °C0,5 °C290 mmRed special liquid57 mmLog in for availability1 * 1 SZT1 020 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 10 CASTM precision thermometer, ASTM 10 CAMARA300170-GA/WCA300170-GA/WC+90...+370 °C2,0 °C290 mmGallium57 mmLog in for availability1 * 1 SZT1 820 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 10 CASTM precision thermometer, ASTM 10 CAMARA300190-GA/WCA300190-GA/WC−6...+400 °C2,0 °C310 mmGallium25 mmLog in for availability1 * 1 SZT1 930 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 12 CASTM precision thermometer, ASTM 12 CAMARA300210-CB/WCA300210-CB/WC−20...+102 °C0,2 °C420 mmBlue special liquidTotalLog in for availability1 * 1 SZT2 040 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 12 CASTM precision thermometer, ASTM 12 CAMARA300210-FL/WCA300210-FL/WC−20...+102 °C0,2 °C420 mmRed special liquidTotalLog in for availability1 * 1 SZT1 660 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 15 CASTM precision thermometer, ASTM 15 CAMARA300260-CB/WCA300260-CB/WC−2...+80 °C0,2 °C395 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 280 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 15 CASTM precision thermometer, ASTM 15 CAMARA300260-FL/WCA300260-FL/WC−2...+80 °C0,2 °C395 mmRed special liquidTotalLog in for availability1 * 1 SZT987 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 16 CASTM precision thermometer, ASTM 16 CAMARA300280-FL/WCA300280-FL/WC+30...+200 °C0,5 °C395 mmRed special liquidTotalLog in for availability1 * 1 SZT1 150 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 17 CASTM precision thermometer, ASTM 17 CAMARA300300-CB/WCA300300-CB/WC+94...+108 °F0,2 °F275 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 110 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 18 CASTM precision thermometer, ASTM 18 CAMARA300320-CB/WCA300320-CB/WC+34...+42 °C0,1 °C275 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 140 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 19 CASTM precision thermometer, ASTM 19 CAMARA300340-CB/WCA300340-CB/WC+49...+57 °C0,1 °C275 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 140 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 21 CASTM precision thermometer, ASTM 21 CAMARA300400-CB/WCA300400-CB/WC+95...+103 °C0,1 °C275 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 290 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 23 CASTM precision thermometer, ASTM 23 CAMARA300420-CB/WCA300420-CB/WC+18...+28 °C0,2 °C212 mmBlue special liquid90 mmLog in for availability1 * 1 SZT1 130 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 23 CASTM precision thermometer, ASTM 23 CAMARA300430-CB/WCA300430-CB/WC+39...+54 °C0,2 °C237 mmBlue special liquid90 mmLog in for availability1 * 1 SZT1 160 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 25 CASTM precision thermometer, ASTM 25 CAMARA300440-CB/WCA300440-CB/WC+95...+105 °C0,2 °C212 mmBlue special liquid90 mmLog in for availability1 * 1 SZT1 300 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 28 CASTM precision thermometer, ASTM 28 CAMARA300468-CB/WCA300468-CB/WC+37...+39 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 550 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 33 CASTM precision thermometer, ASTM 33 CAMARA300500-CB/WCA300500-CB/WC−38...+42 °C0,2 °C420 mmBlue special liquid50 mmLog in for availability1 * 1 SZT1 690 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 34 CASTM precision thermometer, ASTM 34 CAMARA300520-CB/WCA300520-CB/WC+25...+105 °C0,2 °C420 mmBlue special liquid50 mmLog in for availability1 * 1 SZT1 460 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 35 CASTM precision thermometer, ASTM 35 CAMARA300540-CB/WCA300540-CB/WC+90...+170 °C0,2 °C420 mmBlue special liquid50 mmLog in for availability1 * 1 SZT1 630 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 36 CASTM precision thermometer, ASTM 36 CAMARA300560-CB/WCA300560-CB/WC+2...+68 °C0,2 °C405 mmBlue special liquid45 mmLog in for availability1 * 1 SZT1 300 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 37 CASTM precision thermometer, ASTM 37 CAMARA300570-CB/WCA300570-CB/WC−2...+52 °C0,2 °C395 mmBlue special liquid100 mmLog in for availability1 * 1 SZT1 210 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 38 CASTM precision thermometer, ASTM 38 CAMARA300580-CB/WCA300580-CB/WC+24...+78 °C0,2 °C395 mmBlue special liquid100 mmLog in for availability1 * 1 SZT1 260 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 39 CASTM precision thermometer, ASTM 39 CAMARA300590-CB/WCA300590-CB/WC+48...+102 °C0,2 °C395 mmBlue special liquid100 mmLog in for availability1 * 1 SZT1 330 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 40 CASTM precision thermometer, ASTM 40 CAMARA300600-CB/WCA300600-CB/WC+72...+126 °C0,2 °C395 mmBlue special liquid100 mmLog in for availability1 * 1 SZT1 470 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 41 CASTM precision thermometer, ASTM 41 CAMARA300610-CB/WCA300610-CB/WC+98...+152 °C0,2 °C395 mmBlue special liquid100 mmLog in for availability1 * 1 SZT1 470 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 42 CASTM precision thermometer, ASTM 42 CAMARA300620-GA/WCA300620-GA/WC+95...+255 °C0,5 °C395 mmGallium100 mmLog in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 44 CASTM precision thermometer, ASTM 44 CAMARA300634-CB/WCA300634-CB/WC+18,6...+21,4 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 530 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 45 CASTM precision thermometer, ASTM 45 CAMARA300644-CB/WCA300644-CB/WC+23,6…+26,4 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 470 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 46 CASTM precision thermometer, ASTM 46 CAMARA300654-CB/WCA300654-CB/WC+48,6...+51,4 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 590 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 47 CASTM precision thermometer, ASTM 47 CAMARA300664-CB/WCA300664-CB/WC+58,6...+61,4 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 640 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 54 CASTM precision thermometer, ASTM 54 CAMARA300730-CB/WCA300730-CB/WC+20...+100 °C0,2 °C310 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 270 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 57 CASTM precision thermometer, ASTM 57 CAMARA300770-CB/WCA300770-CB/WC−20...+50 °C0,5 °C287 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 220 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 57 CASTM precision thermometer, ASTM 57 CAMARA300770-FL/WCA300770-FL/WC−20...+50 °C0,5 °C287 mmRed special liquidTotalLog in for availability1 * 1 SZT1 010 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 61 CASTM precision thermometer, ASTM 61 CAMARA300820-CB/WCA300820-CB/WC+32...+127 °C0,2 °C380 mmBlue special liquid79 mmLog in for availability1 * 1 SZT1 530 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 62 CASTM precision thermometer, ASTM 62 CAMARA300840-CB/WCA300840-CB/WC−32...+2 °C0,1 °C379 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 650 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 63 CASTM precision thermometer, ASTM 63 CAMARA300860-CB/WCA300860-CB/WC−8...+32 °C0,1 °C379 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 360 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 64 CASTM precision thermometer, ASTM 64 CAMARA300880-CB/WCA300880-CB/WC+25...+55 °C0,1 °C379 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 730 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 65 CASTM precision thermometer, ASTM 65 CAMARA300900-CB/WCA300900-CB/WC+50...+80 °C0,1 °C379 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 750 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 66 CASTM precision thermometer, ASTM 66 CAMARA300920-CB/WCA300920-CB/WC+75...+105 °C0,1 °C379 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 910 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 67 CASTM precision thermometer, ASTM 67 CAMARA300940-CB/WCA300940-CB/WC+95...+155 °C0,2 °C379 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 840 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 68 CASTM precision thermometer, ASTM 68 CAMARA300960-GA/WCA300960-GA/WC+145...+205 °C0,2 °C379 mmGalliumTotalLog in for availability1 * 1 SZT2 660 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 69 CASTM precision thermometer, ASTM 69 CAMARA300980-GA/WCA300980-GA/WC+195...+305 °C0,5 °C379 mmGalliumTotalLog in for availability1 * 1 SZT2 820 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 70 CASTM precision thermometer, ASTM 70 CAMARA301000-GA/WCA301000-GA/WC+295...+405 °C0,5 °C379 mmGalliumTotalLog in for availability1 * 1 SZT3 130 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 73 CASTM precision thermometer, ASTM 73 CAMARA301034-CB/WCA301034-CB/WC−41,4...−38,6 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 640 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 82 FASTM precision thermometer, ASTM 82 FAMARA301140-CB/WCA301140-CB/WC0...+220 °F2,0 °F162 mmBlue special liquid30 mmLog in for availability1 * 1 SZT1 010 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 82 FASTM precision thermometer, ASTM 82 FAMARA301140-FL/WCA301140-FL/WC0...+220 °F2,0 °F162 mmRed special liquid30 mmLog in for availability1 * 1 SZT838 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 83 FASTM precision thermometer, ASTM 83 FAMARA301160-CB/WCA301160-CB/WC+60...+160 °F1,0 °F171 mmBlue special liquid40 mmLog in for availability1 * 1 SZT966 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 83 FASTM precision thermometer, ASTM 83 FAMARA301160-FL/WCA301160-FL/WC+60...+160 °F1,0 °F171 mmRed special liquid40 mmLog in for availability1 * 1 SZT808 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 88 CASTM precision thermometer, ASTM 88 CAMARA301250-FL/WCA301250-FL/WC+10...+200 °C1,0 °C287 mmRed special liquid57 mmLog in for availability1 * 1 SZT1 280 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 88 CASTM precision thermometer, ASTM 88 CAMARA301250-GA/WCA301250-GA/WC+10...+200 °C1,0 °C287 mmGallium57 mmLog in for availability1 * 1 SZT1 560 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 89 CASTM precision thermometer, ASTM 89 CAMARA301270-CB/WCA301270-CB/WC−20...+10 °C0,1 °C370 mmBlue special liquid76 mmLog in for availability1 * 1 SZT1 660 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 90 CASTM precision thermometer, ASTM 90 CAMARA301280-CB/WCA301280-CB/WC0...+30 °C0,1 °C370 mmBlue special liquid76 mmLog in for availability1 * 1 SZT1 390 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 90 CASTM precision thermometer, ASTM 90 CAMARA301280-FL/WCA301280-FL/WC0...+30 °C0,1 °C370 mmRed special liquid76 mmLog in for availability1 * 1 SZT1 100 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 91 CASTM precision thermometer, ASTM 91 CAMARA301290-CB/WCA301290-CB/WC+20...+30 °C0,1 °C370 mmBlue special liquid76 mmLog in for availability1 * 1 SZT1 430 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 99 CASTM precision thermometer, ASTM 99 CAMARA301368-CB/WCA301368-CB/WC−50...+5 °C0,2 °C302 mmBlue special liquid35 mmLog in for availability1 * 1 SZT1 620 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 99 FASTM precision thermometer, ASTM 99 FAMARA301370-CB/WCA301370-CB/WC−58...+41 °C0,5 °F302 mmBlue special liquid35 mmLog in for availability1 * 1 SZT1 620 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 102 CASTM precision thermometer, ASTM 102 CAMARA301400-CB/WCA301400-CB/WC+123,0...+127,0 °C0,2 °C395 mmBlue special liquid100 mmLog in for availability1 * 1 SZT1 490 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 103 CASTM precision thermometer, ASTM 103 CAMARA301410-GA/WCA301410-GA/WC+148,0...+202,0 °C0,2 °C395 mmGallium100 mmLog in for availability1 * 1 SZT1 890 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 104 CASTM precision thermometer, ASTM 104 CAMARA301420-GA/WCA301420-GA/WC+173,0...+227,0 °C0,2 °C395 mmGallium100 mmLog in for availability1 * 1 SZT2 020 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 105 CASTM precision thermometer, ASTM 105 CAMARA301430-GA/WCA301430-GA/WC+198,0...+252,0 °C0,2 °C395 mmGallium100 mmLog in for availability1 * 1 SZT2 070 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 106 CASTM precision thermometer, ASTM 106 CAMARA301440-GA/WCA301440-GA/WC+223,0...+277,0 °C0,2 °C395 mmGallium100 mmLog in for availability1 * 1 SZT2 120 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 107 CASTM precision thermometer, ASTM 107 CAMARA301450-GA/WCA301450-GA/WC+248,0...+302,0 °C0,2 °C395 mmGallium100 mmLog in for availability1 * 1 SZT2 160 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 112 CASTM precision thermometer, ASTM 112 CAMARA301500-CB/WCA301500-CB/WC+4,0...+6,0 °C0,2 °C215 mmBlue special liquidTotalLog in for availability1 * 1 SZT2 460 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 114 CASTM precision thermometer, ASTM 114 CAMARA301530-TOL/WCA301530-TOL/WC−80,0...+20,0 °C0,5 °C300 mmTolueneTotalLog in for availability1 * 1 SZT993 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 120 CASTM precision thermometer, ASTM 120 CAMARA301610-CB/WCA301610-CB/WC+38,6 ...+41,4 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 590 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 121 CASTM precision thermometer, ASTM 121 CAMARA301630-CB/WCA301630-CB/WC+98,6...+101,4 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT2 450 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 122 CASTM precision thermometer, ASTM 122 CAMARA301650-CB/WCA301650-CB/WC−45,0...−35,0 °C0,1 °C300 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 620 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 123 CASTM precision thermometer, ASTM 123 CAMARA301670-CB/WCA301670-CB/WC−35,0...−25,0 °C0,1 °C300 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 540 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 124 CASTM precision thermometer, ASTM 124 CAMARA301690-CB/WCA301690-CB/WC−25,0...−15,0 °C0,1 °C300 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 510 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 125 CASTM precision thermometer, ASTM 125 CAMARA301710-CB/WCA301710-CB/WC−15,0...−5,0 °C0,1 °C300 mmBlue special liquidTotalLog in for availability1 * 1 SZT1 500 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 127 CASTM precision thermometer, ASTM 127 CAMARA301770-CB/WCA301770-CB/WC−21,4...−18,6 °C0,05 °C305 mmBlue special liquidTotalLog in for availability1 * 1 SZT2 000 złSpecifications:
Specifications
Environmentally Preferable
With DKD Calibration Certificate
Product Information
- Catalog #Supplier #RangeDivisionLengthFillingImmersion depthAvailabilityPrice
- ASTM precision thermometer, ASTM 123 CASTM precision thermometer, ASTM 123 CAMARA301670-CBDKD3A301670-CBDKD3−35,0...−25,0 °C0,1 °C300 mmBlue special liquidTotalLog in for availability1 * 1 SZT2 140 złSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 33 CASTM precision thermometer, ASTM 33 CAMARA300500-CBDKD5A300500-CBDKD5−38...+42 °C0,2 °C420 mmBlue special liquid50 mmLog in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable - ASTM precision thermometer, ASTM 124 CASTM precision thermometer, ASTM 124 CAMARA301690-CBDKD3A301690-CBDKD3−25,0...−15,0 °C0,1 °C300 mmBlue special liquidTotalLog in for availability1 * 1 SZTSpecifications:
Specifications
Environmentally Preferable