About this item
The Defender 5000 Series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, powder-coated steel frame and an aluminium load cell. Choice of indicator: D52P unit is ABS plastic and the D52XW indicator is 304 stainless steel with sand blasted surface treatment, which offers protection to IP 68.
- Large backlit display and alpha-numeric keypad with function keys
- Indicator guides user with on-screen prompts
- Multiple application modes and high storage capacity for greater flexibility
- Multiple connectivity options for convenient data transfer and storage
Equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management, and multiple connectivity options - the Defender 5000 is designed to simplify demanding industrial and commercial applications.
The series features common industrial modes and supports a 50000 item library and 1000 user profiles which can be downloaded/uploaded for easy set-up (with SD card.) Also equipped with GLP/GMP output capability with time/date for result traceability.
Weighing results can also be stored in Alibi or regular format, addressing all your data storage needs. Data output via built-in RS232 or optional additional interfaces including second RS232/RS485/USB device, Ethernet, Bluetooth/WiFi, analogue output or discrete I/O with relays, the Defender 5000 indicator can also be used for systems including PLCs.
Results can be read easily on the Defender 5000's large transflective dot matrix display, even in bright sunlight. The SmarText 3.0 interface will guide you through all of the Defender 5000's advanced features. With five soft keys, SmartText 3.0 makes an advanced scale very simple to use, and reduces training time for operators.
Featuring most common applications including percent weighing, parts counting with APW optimisation, checkweighing or checkcounting and dynamic weighing or display hold, the Defender 5000 meets the weighing and measurement needs of practically every industrial application or manufacturing process. Many application modes eliminate the need for long and complicated manual calculations. GMP/GLP with time/date allows for traceability of results.
Models with aluminium load cells, plastic D52P indicator
Product Information
- Catalog #Supplier #ModelWeighing cap.ReadabilityPan/Platform SizePkAvailabilityPrice
- OHAU3044476430444764D52P150RQDX530000 / 60000 g5 / 10 g500×500 mm1Log in for availability1 * 1 SZT5 660 złSupplier Article Number: 30444764Local Article Number: OHAU30444764Specifications:
Specifications
Environmentally Preferable - OHAU3044476230444762D52P60RQDL530000 / 60000 g2 / 5 g400×400 mm1Log in for availability1 * 1 SZT4 250 złSupplier Article Number: 30444762Local Article Number: OHAU30444762Specifications:
Specifications
Environmentally Preferable - OHAU3044476330444763D52P60RQDX530000 / 60000 g2 / 5 g500×500 mm1Log in for availability1 * 1 SZT5 660 złSupplier Article Number: 30444763Local Article Number: OHAU30444763Specifications:
Specifications
Environmentally Preferable - OHAU3042432530424325D52P6RQDR53000 / 6000 g0,2 / 0,5 g305×305 mm1Log in for availability1 * 1 SZT3 745 złSupplier Article Number: 30424325Local Article Number: OHAU30424325Specifications:
Specifications
Environmentally Preferable - OHAU3042432630424326D52P15RQDR56000 / 15000 g0,5 / 1 g305×305 mm1Log in for availability1 * 1 SZT3 745 złSupplier Article Number: 30424326Local Article Number: OHAU30424326Specifications:
Specifications
Environmentally Preferable - OHAU3042432730424327D52P15RQDL56000 / 15000 g0,5 / 1 g400×400 mm1Log in for availability1 * 1 SZT4 250 złSupplier Article Number: 30424327Local Article Number: OHAU30424327Specifications:
Specifications
Environmentally Preferable - OHAU3044476130444761D52P30RQDL515000 / 30000 g1 / 2 g400×400 mm1Log in for availability1 * 1 SZT4 250 złSupplier Article Number: 30444761Local Article Number: OHAU30444761Specifications:
Specifications
Environmentally Preferable
Models with aluminium load cells, stainless steel D52XW indicator (IP 68)
Product Information
- Catalog #Supplier #ModelWeighing cap.ReadabilityPan/Platform SizePkAvailabilityPrice
- OHAU3049821630498216D52XW6RQDR53000 / 6000 g0,2 / 0,5 g305×305 mm1Log in for availability1 * 1 SZT4 930 złSupplier Article Number: 30498216Local Article Number: OHAU30498216Specifications:
Specifications
Environmentally Preferable - OHAU3049821730498217D52XW15RQDR56000 / 15000 g0,5 / 1 g305×305 mm1Log in for availability1 * 1 SZT4 930 złSupplier Article Number: 30498217Local Article Number: OHAU30498217Specifications:
Specifications
Environmentally Preferable - OHAU3049849830498498D52XW15RQDL56000 / 15000 g0,5 / 1 g400×400 mm1Log in for availability1 * 1 SZT5 425 złSupplier Article Number: 30498498Local Article Number: OHAU30498498Specifications:
Specifications
Environmentally Preferable - OHAU3049849930498499D52XW30RQDL515000 / 30000 g1 / 2 g400×400 mm1Log in for availability1 * 1 SZT5 425 złSupplier Article Number: 30498499Local Article Number: OHAU30498499Specifications:
Specifications
Environmentally Preferable - OHAU3049850030498500D52XW60RQDL530000 / 60000 g2 / 5 g400×400 mm1Log in for availability1 * 1 SZT5 425 złSupplier Article Number: 30498500Local Article Number: OHAU30498500Specifications:
Specifications
Environmentally Preferable - OHAU3049850130498501D52XW60RQDX530000 / 60000 g2 / 5 g500×500 mm1Log in for availability1 * 1 SZT6 840 złSupplier Article Number: 30498501Local Article Number: OHAU30498501Specifications:
Specifications
Environmentally Preferable - OHAU3049850230498502D52XW150RQDX530000 / 60000 g5 / 10 g500×500 mm1Log in for availability1 * 1 SZT6 840 złSupplier Article Number: 30498502Local Article Number: OHAU30498502Specifications:
Specifications
Environmentally Preferable
Certified models with aluminium load cells, plastic D52P indicator
Product Information
- Catalog #Supplier #ModelWeighing cap.ReadabilityPan/Platform SizePkAvailabilityPrice
- OHAU3046155830461558D52P6RQDR5-M3000 / 6000 g1 / 2 g305×305 mm1Log in for availability1 * 1 SZT4 215 złSupplier Article Number: 30461558Local Article Number: OHAU30461558Specifications:
Specifications
Environmentally Preferable - OHAU3046155930461559D52P15RQDR5-M6000 / 15000 g2 / 5 g305×305 mm1Log in for availability1 * 1 SZT4 215 złSupplier Article Number: 30461559Local Article Number: OHAU30461559Specifications:
Specifications
Environmentally Preferable - OHAU3046156030461560D52P15RQDL5-M6000 / 15000 g2 / 5 g400×400 mm1Log in for availability1 * 1 SZT4 710 złSupplier Article Number: 30461560Local Article Number: OHAU30461560Specifications:
Specifications
Environmentally Preferable - OHAU3046156130461561D52P30RQDL5-M15000 / 30000 g5 / 10 g400×400 mm1Log in for availability1 * 1 SZT4 710 złSupplier Article Number: 30461561Local Article Number: OHAU30461561Specifications:
Specifications
Environmentally Preferable - OHAU3046156230461562D52P60RQDL5-M30000 / 60000 g10 / 20 g400×400 mm1Log in for availability1 * 1 SZT4 710 złSupplier Article Number: 30461562Local Article Number: OHAU30461562Specifications:
Specifications
Environmentally Preferable - OHAU3046156330461563D52P60RQDX5-M30000 / 60000 g10 / 20 g500×500 mm1Log in for availability1 * 1 SZT6 130 złSupplier Article Number: 30461563Local Article Number: OHAU30461563Specifications:
Specifications
Environmentally Preferable - OHAU3046156430461564D52P150RQDX5-M60000 / 150000 g20 / 50 g500×500 mm1Log in for availability1 * 1 SZT6 130 złSupplier Article Number: 30461564Local Article Number: OHAU30461564Specifications:
Specifications
Environmentally Preferable
Certified models with aluminium load cells, stainless steel D52XW indicator (IP 68)
Product Information
- Catalog #Supplier #ModelWeighing cap.ReadabilityPan/Platform SizePkAvailabilityPrice
- OHAU3046161030461610D52XW6RQDR5-M3000 / 6000 g1 / 2 g305×305 mm1Log in for availability1 * 1 SZT5 865 złSupplier Article Number: 30461610Local Article Number: OHAU30461610Specifications:
Specifications
Environmentally Preferable - OHAU3046161130461611D52XW15RQDR5-M6000 / 15000 g2 / 5 g305×305 mm1Log in for availability1 * 1 SZT5 865 złSupplier Article Number: 30461611Local Article Number: OHAU30461611Specifications:
Specifications
Environmentally Preferable - OHAU3046161230461612D52XW15RQDL5-M6000 / 15000 g2 / 5 g400×400 mm1Log in for availability1 * 1 SZT6 365 złSupplier Article Number: 30461612Local Article Number: OHAU30461612Specifications:
Specifications
Environmentally Preferable - OHAU3046161330461613D52XW30RQDL5-M15000 / 30000 g5 / 10 g400×400 mm1Log in for availability1 * 1 SZT6 365 złSupplier Article Number: 30461613Local Article Number: OHAU30461613Specifications:
Specifications
Environmentally Preferable - OHAU3046161430461614D52XW60RQDL5-M30000 / 60000 g10 / 20 g400×400 mm1Log in for availability1 * 1 SZT6 365 złSupplier Article Number: 30461614Local Article Number: OHAU30461614Specifications:
Specifications
Environmentally Preferable - OHAU3046161530461615D52XW60RQDX5-M30000 / 60000 g10 / 20 g500×500 mm1Log in for availability1 * 1 SZT7 780 złSupplier Article Number: 30461615Local Article Number: OHAU30461615Specifications:
Specifications
Environmentally Preferable - OHAU3046161630461616D52XW150RQDX5-M60000 / 150000 g20 / 50 g500×500 mm1Log in for availability1 * 1 SZT7 780 złSupplier Article Number: 30461616Local Article Number: OHAU30461616Specifications:
Specifications
Environmentally Preferable