Hydrochloric acid 0.1 M HCl (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1789
ADR:8,III
:36.46 g/mol
:GHS05
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Product Information
- Catalog #Supplier #Pack typePk InfoPkAvailabilityPrice
- Supplier Article Number: 35335-1LLocal Article Number: HONC35335-1L
- Supplier Article Number: 35335-6X1LLocal Article Number: HONC35335-6X1L
- Supplier Article Number: 35335-5L-VPLocal Article Number: HONC35335-5L-VP
- Supplier Article Number: 35335-5LLocal Article Number: HONC35335-5L
- Supplier Article Number: 35335-4X5LLocal Article Number: HONC35335-4X5L
- Supplier Article Number: 35335-10L-VPLocal Article Number: HONC35335-10L-VP