Drukuj
Gauze, stainless steel
Dostawca: Thermo Fisher Scientific
42011.VAEA 831 PLN
ALFA42011.VA
Gauze, stainless steel
Stainless steel gauze, 325 mesh woven from 0.04 mm diameter wire, twilled weave, Type 304.
  • Wire cloth in form
  • Store at ambient temperature