Drukuj
Zakrętki do probówek w formacie 96-dołkowym
Dostawca: Micronic
MCRNMP53101EA 2290 PLN
MCRNMP53101 MCRNMP53120 MCRNMP53130 MCRNMP53105
Zakrętki do probówek w formacie 96-dołkowym
Nakrętki Nakrętki do probówek
These screw caps provide a secure closure for long-term sample storage at ultra-low temperatures or for safe sealing during the transport of samples. The caps feature a high quality silicone O-ring around the base to allow the tightest seal possible.
  • Resists chemical solvents
  • Class 7 cleanroom production
  • Free from RNase, DNase and pyrogens
  • Compatible with screw cap tubes in 96-format
Cap height: 14,2 mm
Temperature range: Vapour phase LN₂ to +100 °C