Drukuj
Organic reference standard, Folic acid, (CRM), TraceCERT®, Supelco®
Dostawca: Merck
SUPL74184-50MGEA 0 PLN
SUPL74184-50MG
Organic reference standard, Folic acid, (CRM), TraceCERT®, Supelco®
Certyfikaty: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Uwaga: Storage temperature: 2 - 8° C (refrigerator)