Drukuj
Piece muflowe ze składanymi drzwiczkami, Tmax 1400 °C i sterownikiem B410, L 5/14
Dostawca: Nabertherm
Nabertherm
455-1405EA 40720 PLN
455-1405 455-1613 455-1404 455-1407 455-1616 455-1615 455-1607 455-1618 455-1609 455-1608 455-1397 455-1188 455-1187 455-1399 455-1189 455-1398 455-1400 455-1402 455-1611 455-1191 455-1193 455-1194 455-1186 455-1196
Piece muflowe ze składanymi drzwiczkami, Tmax 1400 °C i sterownikiem B410, L 5/14
Piece i suszarki Piece muflowe i piece do spopielania
These models stand out for their excellent workmanship, advanced and attractive design, and high level of reliability. Heating elements on support tubes radiating freely into the furnace chamber provide particularly short heating times and a maximum temperature of 1400 °C. These muffle furnaces are a good alternative to the LT series when higher application temperatures are required.
  • Heating from two sides
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Only fibre materials which are not classified as carcinogenic according to TRGS 905, Class 1 or 2 are used
  • Dual shell housing for low external temperatures and high stability
  • Fold down door (L) which can be used as a work platform
  • Exhaust air outlet on rear wall of furnace
  • Solid state relays provide low-noise operation
The furnace is equipped with B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The B510 controllers can automatically activate the vent with fan or catalytic converter (not model L 15).
Informacje do zamówienia: All units require a 230 V single-phase power supply. Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of furnace. Manual or automatic gas supply systems are also available. Corresponding models with lift up doors, different Tmax and other controllers are also available. Please search the website for more details.
Uwaga: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.