Order Entry
Polystyrene, analytical standards for GPC
Polystyrene, analytical standards for GPC
Supplier:  Merck
Certificates
undefined
Polystyrene, analytical standards for GPC
Supplier:  Merck
Specifications
  • M-Clarity quality level:
    MQ100
Specifications
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • ~470 Mn, Mw/Mn ~1,11
    ~470 Mn, Mw/Mn ~1,11
    81401-1G
    81401-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 520 zł
    1 520 zł
    1 * 1 g
       1 520 zł
  • ~1020 Mn, Mw/Mn ~1,10
    ~1020 Mn, Mw/Mn ~1,10
    81402-1G
    81402-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 909 zł
    909 zł
    1 * 1 g
       909 zł
  • ~1790 Mn, Mw/Mn ~1,06
    ~1790 Mn, Mw/Mn ~1,06
    81403-1G
    81403-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~4020 Mn, Mw/Mn ~1,05
    ~4020 Mn, Mw/Mn ~1,05
    67302-1G
    67302-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~8950 Mn, Mw/Mn ~1,06
    ~8950 Mn, Mw/Mn ~1,06
    81406-1G
    81406-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 839 zł
    839 zł
    1 * 1 g
       839 zł
  • ~31000 Mn, Mw/Mn ~1,01
    ~31000 Mn, Mw/Mn ~1,01
    81408-1G
    81408-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~31000 Mn, Mw/Mn ~1,01
    ~31000 Mn, Mw/Mn ~1,01
    81408-250MG
    81408-250MG
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 308 zł
    308 zł
    1 * 250 mg
       308 zł
  • ~64000 Mn, Mw/Mn ~1,02
    ~64000 Mn, Mw/Mn ~1,02
    81409-1G
    81409-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~89300 Mn, Mw/Mn ~1,06
    ~89300 Mn, Mw/Mn ~1,06
    81410-1G
    81410-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 939 zł
    939 zł
    1 * 1 g
       939 zł
  • ~151000 Mn, Mw/Mn ~1,02
    ~151000 Mn, Mw/Mn ~1,02
    81411-1G
    81411-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~187000 Mn, Mw/Mn ~1,05
    ~187000 Mn, Mw/Mn ~1,05
    81412-1G
    81412-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~275000 Mn, Mw/Mn ~1,03
    ~275000 Mn, Mw/Mn ~1,03
    81413-1G
    81413-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 873 zł
    873 zł
    1 * 1 g
       873 zł
  • ~930000 Mn, Mw/Mn ~1,03
    ~930000 Mn, Mw/Mn ~1,03
    81416-250MG
    81416-250MG
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 584 zł
    584 zł
    1 * 250 mg
       584 zł
  • Mw 800 (typical)
    Mw 800 (typical)
    327824-1G
    327824-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 558 zł
    558 zł
    1 * 1 g
       558 zł
  • Mw 2500 (typical)
    Mw 2500 (typical)
    327719-1G
    327719-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 419 zł
    419 zł
    1 * 1 g
       419 zł
  • Mw 4000 (typical)
    Mw 4000 (typical)
    327727-1G
    327727-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 525 zł
    525 zł
    1 * 1 g
       525 zł
  • Mw 50000 (typical)
    Mw 50000 (typical)
    330345-1G
    330345-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 555 zł
    555 zł
    1 * 1 g
       555 zł
  • Mw 20000 (typical)
    Mw 20000 (typical)
    327743-1G
    327743-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 432 zł
    432 zł
    1 * 1 g
       432 zł
  • Mw 90000 (typical)
    Mw 90000 (typical)
    327778-1G
    327778-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 467 zł
    467 zł
    1 * 1 g
       467 zł
  • Mw 200000 (typical)
    Mw 200000 (typical)
    327786-1G
    327786-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 549 zł
    549 zł
    1 * 1 g
       549 zł
  • Mw 400000 (typical)
    Mw 400000 (typical)
    327794-1G
    327794-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 555 zł
    555 zł
    1 * 1 g
       555 zł
  • Mw 900000 (typical)
    Mw 900000 (typical)
    327808-1G
    327808-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 529 zł
    529 zł
    1 * 1 g
       529 zł
  • Mw 2000000 (typical)
    Mw 2000000 (typical)
    327816-1G
    327816-1G
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 555 zł
    555 zł
    1 * 1 g
       555 zł