Drukuj
Stablcal reference suspensions
Dostawca: Hach
708-0060EA 974 PLN
708-0060 708-0059
Stablcal reference suspensions
Standardy Wzorce mętności
Ideal for the determination of lucidity/opalescence according to European Pharmacopeia 6.0.
  • Pre-packaged and ready to use
  • Stable for 2 years