May Grunwald's eosin-methylene blue solution (form. RA Lamb), Gurr™ do barwienia preparatów mikroskopowych
351355CEA
208
PLN
351355C
May Grunwald's eosin-methylene blue solution (form. RA Lamb), Gurr™ do barwienia preparatów mikroskopowych
May Grunwald's eosin-methylene blue
Wyniki testów specyfikacyjnych
Suitability for microscopy (blood smear) | Passes test |
Erythrocytes | Red |
Neutrophilic granules | Faintly red |
Eosinophilic granules | Deeper red |
Nuclei | Blue, not violet |
Lymphocyte cytoplasm | Deeper blue |