Drukuj
May Grunwald's eosin-methylene blue solution, Gurr™ do mikroskopii
Dostawca: VWR Chemicals
352065WEA 102 PLN
352065W
May Grunwald's eosin-methylene blue solution, Gurr™ do mikroskopii
May Grunwald's eosin-methylene blue
IVDLogo
Zobacz ceny
Wyniki testów specyfikacyjnych
Suitability for microscopy (blood smear) Passes test
Erythrocytes Pink to brownish
Nuclei Violet
Eosinophilic granules Red to red-brown
Neutrophilic granules Light violet
Lymphocyte cytoplasm Blue